Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC017158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145590.2 | NP_663565.2 | 5, 6 | 641 | ||
GenBank mRNA | AK148236.1 | - | 5, 6 | 632 | ||
AK149645.1 | - | 5, 6 | 620 | |||
AK154566.1 | - | 5, 6 | 607 | |||
AK164488.1 | - | 5, 6 | 621 | |||
AK165287.1 | - | 5, 6 | 620 | |||
AK170611.1 | - | 5, 6 | 620 | |||
AK195588.1 | - | 4, 5 | 205 | |||
BC017158.1 | - | 5, 6 | 623 | 4919869 | ||
BC024080.1 | - | 4, 5 | 251 | |||
BC094040.1 | - | 5, 6 | 605 |