Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195006.1 | NP_001181935.1 | 4, 5 | 431 | ||
NM_145602.3 | NP_663577.1 | 4, 5 | 431 | |||
GenBank mRNA | AK090374.1 | - | 4, 5 | 413 | ||
AK144294.1 | - | 4, 5 | 413 | |||
AK168873.1 | - | 4, 5 | 415 | |||
BC006595.1 | - | 4, 5 | 405 | 3156802 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK129305.1 | - | 726 | Chr. - - [-] on Build |