Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195006.1 | NP_001181935.1 | 15 | 2689 | ||
NM_145602.3 | NP_663577.1 | 14 | 2650 | |||
GenBank mRNA | AK090374.1 | - | 15 | 2672 | ||
AK144294.1 | - | 15 | 2674 | |||
AK168873.1 | - | 15 | 2673 | |||
BC006595.1 | - | 14 | 2624 | 3156802 | ||
BC028336.1 | - | 1 | 1190 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK129305.1 | - | 2984 | Chr. - - [-] on Build |