Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D230025D16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145604.2 | NP_663579.2 | 2 | 345 | ||
GenBank mRNA | AK051965.1 | - | 2 | 331 | ||
AK156120.1 | - | 2 | 347 | |||
AK166937.1 | - | 2 | 328 | |||
BC006874.1 | - | 2 | 228 | 2646515 | ||
BC057919.1 | - | 2 | 334 | 5134302 | ||
BC066141.1 | - | 2 | 266 | |||
BC066826.1 | - | 2 | 266 | |||
CN703950.1 | - | 2 | 345 |