Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145614.4 | NP_663589.3 | 7 | 1368 | ||
GenBank mRNA | AK032124.1 | - | 7 | 1369 | ||
AK043771.1 | - | 7 | 1057 | |||
AK166090.1 | - | 7 | 1138 | |||
AK180711.1 | - | 2 | 239 | |||
AY044265.1 | - | 7 | 805 | |||
BC026680.1 | - | 7 | 1116 | 5059786 | ||
BC069862.1 | - | 7 | 1111 | 5388446 |