Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145619.2 | NP_663594.2 | 7, 8 | 1170 | ||
GenBank mRNA | AF368233.1 | - | 7, 8 | 1170 | ||
AK044223.1 | - | 7, 8 | 1163 | |||
AK088125.1 | - | 7, 8 | 1176 | |||
AK145259.1 | - | 7, 8 | 1204 | |||
AK147868.1 | - | 7, 8 | 1469 | |||
AK170541.1 | - | 7, 8 | 1114 | |||
BC014870.1 | - | 7, 8 | 1110 | 3602116 | ||
BC058754.1 | - | 7, 8 | 1090 | 6476689 | ||
BC062204.1 | - | 7, 8 | 1090 |