Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camkv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145621.2 | NP_663596.1 | 6 | 599 | ||
GenBank mRNA | AK085143.1 | - | 605 | |||
AK147323.1 | - | 6 | 587 | |||
BC017634.1 | - | 6 | 599 | 4527857 | ||
BC022169.1 | - | 5 | 808 | |||
BC086762.1 | - | 5 | 797 | |||
BC111033.1 | - | 6 | 594 | 5368225 |