Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYTH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292018.1 | NP_001278947.1 | 2, 3 | 631 | ||
NM_001292019.1 | NP_001278948.1 | 2, 3 | 197 | |||
NM_004762.3 | NP_004753.1 | 2, 3 | 234 | |||
NM_017456.3 | NP_059430.2 | 2, 3 | 234 | |||
XM_011525475.2 | XP_011523777.1 | 1, 2 | 93 | |||
XM_011525476.2 | XP_011523778.1 | 1, 2 | 93 | |||
XM_011525477.1 | XP_011523779.1 | 1 | 12 | |||
XM_011525478.1 | XP_011523780.1 | 2, 3 | 620 | |||
XM_017025336.1 | XP_016880825.1 | 2, 3 | 304 | |||
XM_017025337.1 | XP_016880826.1 | 2, 3 | 189 | |||
XM_017025338.1 | XP_016880827.1 | 2, 3 | 620 | |||
GenBank mRNA | AK293894.1 | - | 2, 3 | 631 | ||
AK307917.1 | - | 2, 3 | 163 | |||
AK316075.1 | - | 2, 3 | 190 | |||
AK316277.1 | - | 2, 3 | 178 | |||
BC038385.1 | - | 2, 3 | 155 | 4755203 | ||
BC050452.1 | - | 2, 3 | 144 | 6043039 | ||
BX339155.2 | - | 2, 3 | 214 | |||
DA125333.1 | - | 2, 3 | 197 | |||
HY049953.1 | - | 2, 3 | 234 | |||
M85169.1 | - | 2, 3 | 184 |