Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146213.2 | NP_666325.2 | 4 | 633 | ||
GenBank mRNA | AK037191.1 | - | 4 | 556 | ||
BC026374.1 | - | 4 | 633 | 4163362 |