Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146217.4 | NP_666329.2 | 2 | 517 | ||
GenBank mRNA | AK036738.1 | - | 2 | 126 | ||
AK044451.1 | - | 2 | 268 | |||
AK085725.1 | - | 2 | 458 | |||
AK150227.1 | - | 2 | 445 | |||
AK158139.1 | - | 2 | 466 | |||
AK183614.1 | - | 2 | 230 | |||
AK192606.1 | - | 1 | 38 | |||
AY223875.1 | - | 2 | 457 | |||
BC033273.1 | - | 392 | 4196789 | |||
BM943271.1 | - | 2 | 162 |