Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146254.4 | NP_666366.4 | 2 | 231 | ||
GenBank mRNA | BC028975.1 | - | 2 | 156 | 5363117 | |
CF742041.1 | - | 2 | 488 |