Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146254.4 | NP_666366.4 | 11 | 1618 | ||
GenBank mRNA | AK030997.1 | - | 4 | 3303 | ||
BC028975.1 | - | 11 | 1543 | 5363117 | ||
BC034130.1 | - | 3 | 598 | 4920514 |