Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpdl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146256.3 | NP_666368.1 | 1 | 797 | ||
GenBank mRNA | AK043902.1 | - | 1 | 778 | ||
AK049217.1 | - | 1 | 777 | |||
AK078929.1 | - | 1 | 798 | |||
BC034099.1 | - | 1 | 612 | 4489103 |