Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly6g5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148939.2 | NP_683741.1 | 3 | 233 | ||
GenBank mRNA | AJ315553.1 | - | 3 | 233 | ||
AJ315554.1 | - | 2 | 329 | |||
AK151902.1 | - | 2 | 270 | |||
BC131964.1 | - | 3 | 261 | 40130241 | ||
BC131966.1 | - | 3 | 261 | 40130243 | ||
BC145150.1 | - | 2 | 357 | |||
HE864438.1 | - | 3 | 249 | |||
HE864439.1 | - | 2 | 345 |