Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153105.7 | NP_694745.1 | 4 | 833 | ||
GenBank mRNA | AK032743.1 | - | 4 | 766 | ||
AK143945.1 | - | 4 | 835 | |||
BC114989.1 | - | 4 | 766 | 40056185 | ||
BC115827.1 | - | 4 | 766 | 40056184 |