Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153171.4 | NP_694811.1 | 6 | 529 | ||
GenBank mRNA | AF498319.1 | - | 6 | 578 | ||
AK043268.1 | - | 6 | 529 | |||
BC049624.1 | - | 6 | 447 | 6585574 |