Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ago2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153178.4 | NP_694818.3 | 2 | 274 | ||
GenBank mRNA | AB081472.1 | - | 2 | 136 | ||
BC096465.1 | - | 2 | 179 | |||
BC128379.1 | - | 2 | 127 | 40111876 | ||
BC129922.1 | - | 2 | 127 | 40111875 | ||
BY732637.1 | - | 2 | 229 | |||
CJ056675.1 | - | 2 | 266 |