Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB33A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004794.2 | NP_004785.1 | 1, 2 | 515 | ||
GenBank mRNA | AF498959.1 | - | 1, 2 | 251 | ||
BC001157.1 | - | 1, 2 | 515 | 3510745 | ||
BC009996.2 | - | 1, 2 | 515 | 4129324 | ||
BT007038.1 | - | 1, 2 | 251 | |||
D14889.1 | - | 1, 2 | 688 |