Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256196.1 | NP_001243125.1 | 16 | 2456 | ||
NM_001654.4 | NP_001645.1 | 16 | 2447 | |||
XM_006724529.2 | XP_006724592.1 | 16 | 2383 | |||
XM_011543906.2 | XP_011542208.1 | 16 | 3773 | |||
XM_011543908.2 | XP_011542210.1 | 16 | 3757 | |||
XM_011543909.2 | XP_011542211.1 | 11 | 1748 | |||
GenBank mRNA | AB208831.1 | - | 12 | 5514 | ||
AF052136.1 | - | 6 | 1154 | |||
AK130043.1 | - | 15 | 1743 | |||
BC002466.2 | - | 16 | 2327 | 3347509 | ||
BC007514.2 | - | 16 | 2334 | 2966604 | ||
X04790.1 | - | 16 | 2430 |