Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172147.1 | NP_001165618.1 | 6 | 998 | ||
NM_001172148.1 | NP_001165619.1 | 5 | 916 | |||
NM_001290630.1 | NP_001277559.1 | 6 | 988 | |||
NM_153586.2 | NP_705814.1 | 5 | 916 | |||
GenBank mRNA | AK040213.1 | - | 5 | 856 | ||
AK080041.1 | - | 3 | 682 | |||
AK084514.1 | - | 5 | 851 | |||
AK084629.1 | - | 5 | 851 | |||
AK141075.1 | - | 6 | 923 | |||
AK163147.1 | - | 4 | 836 | |||
AK164496.1 | - | 6 | 946 | |||
AK166005.1 | - | 5 | 851 | |||
BC037024.1 | - | 5 | 883 | 5368185 |