Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcmtd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291211.1 | NP_001278140.1 | 6 | 938 | ||
NM_153594.3 | NP_705822.1 | 6 | 934 | |||
GenBank mRNA | AB182640.1 | - | 6 | 879 | ||
AK031335.1 | - | 6 | 889 | |||
AK079927.1 | - | 3 | 210 | |||
AK088800.1 | - | 6 | 873 | |||
AK146080.1 | - | 6 | 881 | |||
AK204607.1 | - | 2 | 105 | |||
BC024791.1 | - | 1 | 28 | |||
BC031360.1 | - | 5 | 751 | 5359242 | ||
BC040385.1 | - | 6 | 885 | 3499499 |