Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcmtd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291211.1 | NP_001278140.1 | 7 | 3345 | ||
NM_153594.3 | NP_705822.1 | 7 | 3341 | |||
GenBank mRNA | AB182640.1 | - | 7 | 3286 | ||
AK088800.1 | - | 7 | 3280 | |||
AK146080.1 | - | 7 | 3288 | |||
BC040385.1 | - | 7 | 3292 | 3499499 |