Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx19b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190786.1 | NP_001177715.1 | 12 | 1931 | ||
NM_001190800.1 | NP_001177729.1 | 12 | 1969 | |||
NM_172284.5 | NP_758488.1 | 12 | 1974 | |||
GenBank mRNA | AK033235.1 | - | 12 | 1932 | ||
AK147499.1 | - | 12 | 1969 | |||
BC025594.1 | - | 12 | 1934 | 5342859 |