Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc36a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172289.4 | NP_758493.2 | 10 | 1107 | ||
GenBank mRNA | AF453746.1 | - | 11 | 1106 | ||
AK032074.1 | - | 10 | 1108 | |||
AK052277.1 | - | 10 | 1108 | |||
AK150167.1 | - | 10 | 1139 | |||
BC115964.1 | - | 10 | 1108 | 40057255 |