Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mab21l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172295.4 | NP_758499.3 | 7 | 2310 | ||
GenBank mRNA | AK029040.1 | - | 7 | 2049 | ||
AK161057.1 | - | 7 | 2227 | |||
AK166928.1 | - | 2060 | ||||
AK172389.1 | - | 7 | 2259 | |||
BC037703.1 | - | 7 | 2247 | 4166596 |