Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143995.2 | NP_001137467.1 | 2 | 366 | ||
NM_001307951.1 | NP_001294880.1 | 2 | 399 | |||
NM_004811.2 | NP_004802.1 | 2 | 303 | |||
XM_006718750.1 | XP_006718813.1 | 2 | 399 | |||
XM_011545388.2 | XP_011543690.1 | 2 | 1070 | |||
XM_011545390.2 | XP_011543692.1 | 2 | 1380 | |||
XM_011545391.2 | XP_011543693.1 | 2 | 346 | |||
XM_011545393.2 | XP_011543695.1 | 2 | 346 | |||
XM_011545394.2 | XP_011543696.1 | 2 | 269 | |||
XM_017018579.1 | XP_016874068.1 | 2 | 346 | |||
GenBank mRNA | AF062075.1 | - | 2 | 251 | ||
AK223165.1 | - | 2 | 242 | |||
AK299264.1 | - | 2 | 380 | |||
AK300955.1 | - | 2 | 293 | |||
AK301093.1 | - | 2 | 242 | |||
AK313306.1 | - | 2 | 236 | |||
BC019035.2 | - | 2 | 225 | 4575069 | ||
BP261314.1 | - | 2 | 303 | |||
CR536531.1 | - | 1 | 158 | |||
DA363664.1 | - | 2 | 400 |