Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpsf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164272.1 | NP_001157744.1 | 3 | 234 | ||
NM_172302.3 | NP_758506.3 | 3 | 234 | |||
GenBank mRNA | AK044306.1 | - | 3 | 213 | ||
AK053375.1 | - | 3 | 213 | |||
AK088603.1 | - | 3 | 212 | |||
AK165330.1 | - | 3 | 212 | |||
AK188327.1 | - | 2 | 102 | |||
AK189206.1 | - | 2 | 102 | |||
BC017519.1 | - | 2 | 154 | |||
BC038812.1 | - | 2 | 164 | 4506179 | ||
BY076191.1 | - | 3 | 235 |