Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpsf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164272.1 | NP_001157744.1 | 10 | 2481 | ||
NM_172302.3 | NP_758506.3 | 10 | 2508 | |||
GenBank mRNA | AK053375.1 | - | 11 | 2553 | ||
AK082789.1 | - | 1 | 506 | |||
AK088603.1 | - | 10 | 2486 | |||
BC017519.1 | - | 9 | 2399 | |||
BC026433.1 | - | 3 | 1129 | |||
BC038812.1 | - | 9 | 2411 | 4506179 |