Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahsa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290654.1 | NP_001277583.1 | 3 | 491 | ||
NM_001290655.1 | NP_001277584.1 | 3 | 496 | |||
NM_172391.4 | NP_765979.3 | 3 | 496 | |||
GenBank mRNA | AK033696.1 | - | 3 | 404 | ||
AK035941.1 | - | 3 | 269 | |||
AK093945.1 | - | 3 | 403 | |||
AK137116.1 | - | 3 | 407 | |||
AK154196.1 | - | 3 | 407 | |||
AK155265.1 | - | 3 | 407 | |||
AK186357.1 | - | 3 | 248 | |||
BC038397.1 | - | 3 | 382 | 3676293 | ||
BC052829.1 | - | 3 | 414 | 30040034 | ||
BC055873.1 | - | 3 | 358 | |||
BC064012.1 | - | 3 | 316 | 30135289 | ||
BY023007.1 | - | 3 | 381 | |||
CJ147821.1 | - | 3 | 497 |