Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tufm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163713.1 | NP_001157185.1 | 5 | 712 | ||
NM_172745.3 | NP_766333.1 | 5 | 712 | |||
GenBank mRNA | AK075857.1 | - | 5 | 636 | ||
AK084724.1 | - | 5 | 693 | |||
AK152858.1 | - | 5 | 707 | |||
AK153135.1 | - | 5 | 707 | |||
AK180451.1 | - | 1 | 83 | |||
BC060959.1 | - | 5 | 689 | 30379757 | ||
BC100596.1 | - | 5 | 705 | 6762419 |