Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adprhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172750.3 | NP_766338.1 | 7 | 1257 | ||
GenBank mRNA | AJ427360.1 | - | 7 | 1234 | ||
AK081786.1 | - | 7 | 1091 | |||
AK084501.1 | - | 7 | 1258 | |||
BC063759.1 | - | 7 | 1247 | 30134000 |