Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172756.2 | NP_766344.2 | 5 | 955 | ||
GenBank mRNA | AK049395.1 | - | 3 | 1262 | ||
AK078830.1 | - | 4 | 1185 | |||
AK080768.1 | - | 3 | 1268 | |||
AK083552.1 | - | 3 | 1254 | |||
AK087444.1 | - | 3 | 1272 | |||
BC030436.1 | - | 5 | 894 | |||
BC094316.1 | - | 5 | 912 | |||
BC145659.1 | - | 1 | 178 | |||
CJ046325.1 | - | 4 | 410 | |||
EU184014.1 | - | 5 | 931 |