Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heatr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172757.3 | NP_766345.3 | 15 | 2665 | ||
GenBank mRNA | AK052635.1 | - | 15 | 2692 | ||
AK134510.1 | - | 15 | 2670 | |||
AK140522.1 | - | 15 | 2683 | |||
AK163533.1 | - | 2 | 741 | |||
AK163715.1 | - | 15 | 3428 | |||
AK165541.1 | - | 15 | 2560 | |||
BC030373.1 | - | 11 | 2010 | |||
BC048074.1 | - | 4 | 1383 | |||
BC065060.1 | - | 15 | 2665 | 6402217 |