Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heatr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172757.3 | NP_766345.3 | 15 | 2136 | ||
GenBank mRNA | AK047782.1 | - | 15 | 2167 | ||
AK052635.1 | - | 15 | 2163 | |||
AK134510.1 | - | 15 | 2141 | |||
AK136424.1 | - | 15 | 2146 | |||
AK140522.1 | - | 15 | 2154 | |||
AK163533.1 | - | 2 | 212 | |||
AK163715.1 | - | 15 | 2899 | |||
AK165541.1 | - | 15 | 2030 | |||
BC030373.1 | - | 11 | 1481 | |||
BC048074.1 | - | 4 | 854 | |||
BC065060.1 | - | 15 | 2136 | 6402217 |