Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfrkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172766.3 | NP_766354.2 | 4 | 482 | ||
GenBank mRNA | AK036381.1 | - | 1 | 1941 | ||
AK041206.1 | - | 4 | 478 | |||
BC030051.1 | - | 1 | 33 | |||
BC033595.1 | - | 4 | 472 | 4947357 | ||
BC059861.1 | - | 4 | 461 | |||
BC060076.1 | - | 4 | 469 |