Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gramd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172768.1 | NP_766356.1 | 12 | 1685 | ||
GenBank mRNA | AK028277.1 | - | 13 | 1580 | ||
AK033156.1 | - | 12 | 1685 | |||
AK122465.1 | - | 11 | 1645 | |||
BC030911.1 | - | 3 | 430 | |||
BC070451.1 | - | 11 | 1519 | 30616509 |