Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172780.3 | NP_766368.2 | 3, 4 | 638 | ||
GenBank mRNA | AK032326.1 | - | 3, 4 | 619 | ||
AK122232.1 | - | 3, 4 | 609 | |||
BC130221.1 | - | 3, 4 | 623 | 40141414 |