Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3rf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172788.3 | NP_766376.2 | 10 | 3665 | ||
GenBank mRNA | BC057304.1 | - | 10 | 2921 | 6830567 |