Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btnl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172793.2 | NP_766381.1 | 10 | 2308 | ||
GenBank mRNA | AK080918.1 | - | 10 | 2290 | ||
AK084528.1 | - | 10 | 2309 |