Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp454 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172794.2 | NP_766382.2 | 6 | 1062 | ||
GenBank mRNA | AK046117.1 | - | 3 | 811 | ||
BC049793.1 | - | 6 | 1062 | 6510760 |