Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sarm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168521.1 | NP_001161993.1 | 9 | 2764 | ||
NM_172795.3 | NP_766383.2 | 9 | 2644 | |||
GenBank mRNA | AK044113.1 | - | 4 | 981 | ||
AK172964.1 | - | 8 | 2907 | |||
BC052098.1 | - | 9 | 2290 | |||
BC058534.1 | - | 9 | 2591 | 6834056 | ||
BC080850.1 | - | 9 | 2644 | 6849138 |