Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172799.4 | NP_766387.2 | 7 | 791 | ||
GenBank mRNA | AK077033.1 | - | 6 | 1278 | ||
AM690749.1 | - | 7 | 791 | |||
BC132203.1 | - | 6 | 919 | 40130480 | ||
BC138058.1 | - | 6 | 919 | 8861077 | ||
BC145130.1 | - | 6 | 920 | 9053678 | ||
BC145131.1 | - | 6 | 919 | 9053679 |