Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172800.2 | NP_766388.2 | 7, 8 | 1011 | ||
GenBank mRNA | AK030591.1 | - | 2109 | |||
AK052040.1 | - | 7, 8 | 1036 | |||
AK129379.1 | - | 3, 4 | 392 | |||
AY351699.1 | - | 7, 8 | 1011 | |||
BC038036.1 | - | 7, 8 | 1289 |