Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lax1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159649.1 | NP_001153121.1 | 6 | 1014 | ||
NM_172842.3 | NP_766430.3 | 6 | 916 | |||
GenBank mRNA | AK040834.1 | - | 6 | 824 | ||
AK088503.1 | - | 6 | 767 | |||
AY090785.1 | - | 5 | 642 | |||
BC042765.1 | - | 6 | 916 | 4188712 |