Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276336.1 | NP_001263265.1 | 4 | 433 | ||
NM_001276337.1 | NP_001263266.1 | 3 | 298 | |||
NM_001282958.1 | NP_001269887.1 | 4 | 433 | |||
NM_004846.3 | NP_004837.1 | 4 | 433 | |||
XM_005246975.3 | XP_005247032.1 | 3 | 249 | |||
XM_011512206.2 | XP_011510508.1 | 4 | 708 | |||
XM_017005377.1 | XP_016860866.1 | 3 | 249 | |||
GenBank mRNA | AB209519.1 | - | 4 | 337 | ||
AF038957.1 | - | 4 | 326 | |||
AF047695.1 | - | 4 | 324 | |||
AF068117.1 | - | 4 | 309 | |||
AK022518.1 | - | 4 | 331 | |||
AK303802.1 | - | 3 | 196 | |||
BC005392.1 | - | 4 | 331 | 3997986 | ||
BC005874.2 | - | 4 | 353 | 2961437 | ||
BC021226.2 | - | 4 | 297 | 4302323 | ||
BC021690.1 | - | 4 | 335 | 3961350 | ||
BG913475.1 | - | 4 | 334 | |||
CB994873.1 | - | 401 | ||||
CD684451.1 | - | 3 | 221 | |||
DA408433.1 | - | 2 | 53 | |||
DA992544.1 | - | 4 | 433 |