Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfml2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172854.2 | NP_766442.1 | 6 | 1135 | ||
GenBank mRNA | AK030069.1 | - | 958 | |||
AK035313.1 | - | 6 | 1136 | |||
AK220202.1 | - | 5 | 934 | |||
BC137871.1 | - | 6 | 1063 | 8860890 |