Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pramef8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172877.2 | NP_766465.1 | 5 | 3025 | ||
GenBank mRNA | AK029142.1 | - | 5 | 3026 | ||
BC056931.1 | - | 4 | 2399 | 6816019 |