Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172883.2 | NP_766471.1 | 10 | 1878 | ||
GenBank mRNA | AK029021.1 | - | 10 | 1878 | ||
AK087226.1 | - | 10 | 2090 | |||
AK171069.1 | - | 9 | 4510 | |||
BC094512.1 | - | 8 | 1484 | 2616131 |