Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scml4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172938.3 | NP_766526.3 | 9 | 2259 | ||
GenBank mRNA | AF397912.1 | - | 2 | 797 | ||
AK034171.1 | - | 9 | 2098 | |||
AK040597.1 | - | 4 | 1907 | |||
AK043500.1 | - | 4 | 1790 | |||
AK047972.1 | - | 7 | 1685 | |||
AK141826.1 | - | 4 | 1284 | |||
AK172160.1 | - | 5 | 1322 | |||
AK220309.1 | - | 7 | 1643 | |||
BC024570.1 | - | 4 | 1225 | |||
BC039567.1 | - | 2 | 782 | |||
BC043310.1 | - | 9 | 2064 | 5067233 |