Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172945.2 | NP_766533.2 | 16 | 2843 | ||
GenBank mRNA | AK081149.1 | - | 1973 | |||
BC022693.1 | - | 5 | 1389 | |||
BC039786.1 | - | 5 | 1807 | |||
BC056928.1 | - | 9 | 2329 | |||
BC071189.1 | - | 10 | 1929 | |||
BC098207.1 | - | 7 | 2139 |